3'-Aminoacetanilide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-(3-Aminophenyl)acetamide, 3-Acetamidoaniline, N-Acetyl-1,3-phenylenediamine, m-Aminoacetanilide

XBX00314.250 XBX00314.1
MAYBXBX00314.250EA 0 EUR
MAYBXBX00314.250 MAYBXBX00314.1
3'-Aminoacetanilide
3'-Aminoacetanilide
Formule: C₈H₁₀N₂O
MW: 150.18 g/mol
Kookpunt: 388
Smeltpunt: 84…86 °C
MDL Nummer: MFCD00025229
CAS nummer: 102-28-3
EINECS: 203-021-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR