4'-Fluoroacetanilide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-Fluoroacetanilide, N-(4-Fluorophenyl)acetamide

DP01919.1 DP01919.10 DP01919.50
MAYBDP01919.1EA 0 EUR
MAYBDP01919.1 MAYBDP01919.10 MAYBDP01919.50
4'-Fluoroacetanilide
4'-Fluoroacetanilide
Formule: C₈H₈FNO
MW: 153.16 g/mol
Kookpunt: 310
Smeltpunt: 154
MDL Nummer: MFCD00016335
CAS nummer: 351-83-7
EINECS: 206-515-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR