Phenylacetylene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
E0196-25ML E0196-500ML E0196-100ML
TCIAE0196-25MLEA 85.8 EUR
TCIAE0196-25ML TCIAE0196-500ML TCIAE0196-100ML
Phenylacetylene ≥98.0% (by GC)
Phenylacetylene
Formule: C₈H₆
Kookpunt: 142°C
Dichtheid: 1,1 g/cm³ (20 °C)
Vlampunt: 27°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008570
CAS nummer: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III
Merck Index: 14,03861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=120e75b432082e6e8e43dc3bc4be3eafe0de7e72&vid=120e75b432082e6e8e43dc3bc4be3eafe0de7e72&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %