Acetaal ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D0455-25ML D0455-500ML
TCIAD0455-25MLEA 17.6 EUR
TCIAD0455-25ML TCIAD0455-500ML
Acetaal ≥98.0% (by GC)
Acetaal
Formule: C₆H₁₄O₂
Kookpunt: 103°C
Smeltpunt: -100°C
Dichtheid: 0,83 g/cm³ (20 °C)
Vlampunt: -21°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009243
CAS nummer: 105-57-7
EINECS: 203-310-6
UN: 1088
ADR: 3,II
Merck Index: 14,00038
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0cec5ae42445f700d4e82d619c526b4081a4e5f&vid=b0cec5ae42445f700d4e82d619c526b4081a4e5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR