2-Methoxy-5-nitrophenol ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0498-25G
N0498-500G
TCIAN0498-25GEA
41.8
EUR
TCIAN0498-25G
TCIAN0498-500G
2-Methoxy-5-nitrophenol ≥98.0% (by titrimetric analysis)
2-Methoxy-5-nitrophenol
Formule:
C₇H₇NO₄ Kookpunt: 112°C Smeltpunt: 103…106 °C Dichtheid: 1 g/cm³ (20 °C) |
MDL Nummer:
MFCD00015561 CAS nummer: 636-93-1 EINECS: 211-269-0 |
|
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 103.0 to 106.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...