(±)-1-Phenyl-1,2-ethanediol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P0686-25G P0686-250G
TCIAP0686-25GEA 83.6 EUR
TCIAP0686-25G TCIAP0686-250G
(±)-1-Phenyl-1,2-ethanediol ≥98.0% (by GC)
(±)-1-Phenyl-1,2-ethanediol
Formule: C₆H₅CH(OH)CH₂OH
Kookpunt: 82°C
Smeltpunt: 61…64 °C
MDL Nummer: MFCD00003546
CAS nummer: 93-56-1
EINECS: 202-258-1
Merck Index: 14,08861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eac30ac4d73a63ad627939278dd241035186523f&vid=eac30ac4d73a63ad627939278dd241035186523f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 68.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR