(±)-γ-Nonalactone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: gamma-Nonalactone, (±)-Nonan-4-olide, gamma-Nonanolactone, 5-Pentyldihydrofuran-2(3H)-one, gamma-Pelargonolactone

N0285-25ML N0285-500ML
TCIAN0285-25MLEA 17.6 EUR
TCIAN0285-25ML TCIAN0285-500ML
(±)-γ-Nonalactone ≥98.0% (by GC)
(±)-γ-Nonalactone
Formule: C₉H₁₆O₂
Kookpunt: 243°C
Dichtheid: 0,97 g/cm³ (20 °C)
Vlampunt: 132°C
MDL Nummer: MFCD00005403
CAS nummer: 104-61-0
EINECS: 203-219-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3137bfbe122d40281d0ab0747a9fefa26f3b7e45&vid=3137bfbe122d40281d0ab0747a9fefa26f3b7e45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR