(±)-1,2-Epithiopropane ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar


Stabilisatie: Stabilised with butyl mercaptan

P0321-25ML P0321-5ML
TCIAP0321-25MLEA 163 EUR
TCIAP0321-25ML TCIAP0321-5ML
(±)-1,2-Epithiopropane ≥98.0% (by GC) stabilised
(±)-1,2-Epithiopropane
Formule: C₃H₆S
Kookpunt: 75°C
Smeltpunt: –91 °C
Dichtheid: 0,964 g/cm³ (0 °C)
Vlampunt: 10°C
MDL Nummer: MFCD00005160
CAS nummer: 1072-43-1
EINECS: 214-007-3
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3decdcb11393511e33899588c6ff01afa8bd944&vid=f3decdcb11393511e33899588c6ff01afa8bd944&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR