4-Chlorostyrene ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-tert-butylpyrocatechol (TBC)

C0290-10ML C0290-25ML
TCIAC0290-10MLEA 24.2 EUR
TCIAC0290-10ML TCIAC0290-25ML
4-Chlorostyrene ≥98.0% (by GC) stabilised
4-Chlorostyrene
Formule: C₈H₇Cl
Kookpunt: 192 °C (1013 hPa)
Dichtheid: 1,155 g/cm³ (20 °C)
Vlampunt: 64°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000632
CAS nummer: 1073-67-2
EINECS: 214-028-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c797076ca4863952e3a5d5ac7540644bb8e8b54&vid=2c797076ca4863952e3a5d5ac7540644bb8e8b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR