Osmiumtetraoxide 4% in aqueous solution, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
M-Clarity™ quality level = MQ100
75632-10ML 75632-5ML
75632-10MLEA 470 EUR
75632-10ML 75632-5ML
Osmiumtetraoxide 4% in aqueous solution, Sigma-Aldrich®
Osmiumtetraoxide
Osmium tetroxide is also referred to as osmium and is mainly used as a secondary fixative of tissues. The molecule reacts with lipid moieties and is responsible for the oxidation of unsaturated bonds of fatty acids. This reaction adds density and contrast to biological samples. In normal tissues, presence of osmium results in intense black staining which can be viewed by normal eye. In addition, it can be used as a mordant (a molecule which combines with stain or dye at later stages) in electron microscopy, thereby enhancing lead staining.
Osmium tetroxide solution has been used as a fixative in scanning and transmission electron microscopy.
Formule: O₄Os
MW: 254.23 g/mol
Kookpunt: 130
Smeltpunt: 40
Dichtheid: 1,0 g/ml at 20 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1 II
Merck Index: 13,06962
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d1e1f464bd2f74057d356545f23ca14bd1b5c4&vid=28b411820c6daad9ea39b4cb325bf7c929475661&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 4.0% OsO₄ in water