(±)-Camphor-10-sulphonic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: DL-Camphor-10-sulphonic acid, (±)-2-Oxobornane-10-sulphonic acid, (7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid

C0016-25G C0016-500G C0016-100G
TCIAC0016-25GEA 17.6 EUR
TCIAC0016-25G TCIAC0016-500G TCIAC0016-100G
(±)-Camphor-10-sulphonic acid ≥98.0% (by titrimetric analysis)
(±)-Camphor-10-sulphonic acid
Formule: C₁₀H₁₆O₄S
MW: 232.3
Smeltpunt: 202°C
MDL Nummer: MFCD00074827
CAS nummer: 5872-08-2
EINECS: 227-527-0
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f52a279cc7edf14d4135d26bcecea3242bfab57&vid=5f52a279cc7edf14d4135d26bcecea3242bfab57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -1.0 to +1.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR