L(-)-Camphorsulphonic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0972-25G
C0972-500G
C0972-100G
TCIAC0972-25GEA
22
EUR
TCIAC0972-25G
TCIAC0972-500G
TCIAC0972-100G
L(-)-Camphorsulphonic acid ≥98.0% (by titrimetric analysis)
L(-)-Camphorsulphonic acid
Formule:
C₁₀H₁₆O₄S Kookpunt: 375 °C (1013 hPa) Smeltpunt: 195°C Dichtheid: 1 g/cm³ (20 °C) |
MDL Nummer:
MFCD00064158 CAS nummer: 35963-20-3 EINECS: 252-817-9 UN: 3261 ADR: 8,III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -20.0 to -23.0 deg (C=5, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...