1,1,1-Tris(hydroxymethyl)ethane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol

T0479-25G T0479-500G
TCIAT0479-25GEA 17.6 EUR
TCIAT0479-25G TCIAT0479-500G
1,1,1-Tris(hydroxymethyl)ethane ≥98.0% (by GC)
1,1,1-Tris(hydroxymethyl)ethane
Formule: C₅H₁₂O₃
MW: 120.15 g/mol
Kookpunt: 137°C
MDL Nummer: MFCD00004687
CAS nummer: 77-85-0
EINECS: 201-063-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=899ebcfc7594360342963c7d8de7af3bc37cb781&vid=899ebcfc7594360342963c7d8de7af3bc37cb781&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 203.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR