Octyl acetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A0042-25ML A0042-500ML
TCIAA0042-25MLEA 17.6 EUR
TCIAA0042-25ML TCIAA0042-500ML
Octyl acetate ≥98.0% (by GC)
Octyl acetate
Formule: C₁₀H₂₀O₂
Kookpunt: 210°C
Dichtheid: 0,885 g/cm³ (0 °C)
Vlampunt: 95°C
MDL Nummer: MFCD00009562
CAS nummer: 112-14-1
EINECS: 203-939-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fb9d5807b1d67cb3c1e42b89204f66134d09de&vid=5f4e5317e303701d44f625e6818c40a68d1fb98b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 98.0 %