1,8-Diiodooctane ≥95.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with copper chip
D2158-25G
D2158-5G
TCIAD2158-25GEA
130
EUR
TCIAD2158-25G
TCIAD2158-5G
1,8-Diiodooctane ≥95.0% (by GC) stabilised
1,8-Diiodooctane
Formule:
C₈H₁₆I₂ Kookpunt: 144°C Smeltpunt: 16…21 °C Dichtheid: 1,8375 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001106 CAS nummer: 24772-63-2 EINECS: 246-456-6 |
|
Specification Test Results
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...