Propargyl chloride ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
P0810-25ML P0810-5ML
TCIAP0810-25MLEA 46.2 EUR
TCIAP0810-25ML TCIAP0810-5ML
Propargyl chloride ≥97.0% (by GC)
Propargyl chloride
Formule: C₃H₃Cl
Kookpunt: 57°C
Smeltpunt: –77 °C
Dichtheid: 1,0454 g/cm³ (5 °C)
Vlampunt: 19°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000980
CAS nummer: 624-65-7
EINECS: 210-856-9
UN: 3286
ADR: 3,II
Merck Index: 14,07810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a03176c6f90f6a16f6843f5767f6ace9cfc7271&vid=4a03176c6f90f6a16f6843f5767f6ace9cfc7271&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %