1H,1H,2H,2H-Tridecafluoro-1-octanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol
T2528-25G T2528-5G
TCIAT2528-25GEA 49.5 EUR
TCIAT2528-25G TCIAT2528-5G
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥98.0% (by GC)
1H,1H,2H,2H-Tridecafluoro-1-octanol
Formule: C₈H₅F₁₃O
Kookpunt: 87°C
Vlampunt: 91°C
MDL Nummer: MFCD00042143
CAS nummer: 647-42-7
EINECS: 211-477-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbc7d11d291175188f71c73dbea523d3153dffa2&vid=dbc7d11d291175188f71c73dbea523d3153dffa2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %