Potassium cyclopropyltrifluoroborate ≥90%, tech.
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cyclopropyltrifluoroboric acid potassium salt
H60673.03
H60673.06
H60673.03EA
81.4
EUR
H60673.03
H60673.06
Potassium cyclopropyltrifluoroborate ≥90%, tech.
Potassium cyclopropyltrifluoroborate
Formule:
C₃H₅BF₃K Smeltpunt: 348…350 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09265031 CAS nummer: 1065010-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...