2,5-Dihydroxybenzoic acid 99,0 GC, T Matrix for MALDI-TOF/MS
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,5-Dihydroxybenzoic Acid, DHB, Hydroquinonecarboxylic acid, Gentisic acid, 2,5-DHBA
D2933-5G
D2933-1G
TCIAD2933-5GEA
71.5
EUR
TCIAD2933-5G
TCIAD2933-1G
2,5-Dihydroxybenzoic acid 99,0 GC, T Matrix for MALDI-TOF/MS
2,5-Dihydroxybenzoic acid
Formule:
C₇H₆O₄ MW: 154.12 g/mol Kookpunt: 339 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002460 CAS nummer: 490-79-9 EINECS: 207-718-5 Merck Index: 14,04398 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...