4'-Methylbiphenyl-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2377-25G T2377-5G
TCIAT2377-25GEA 48.4 EUR
TCIAT2377-25G TCIAT2377-5G
4'-Methylbiphenyl-2-carboxylic acid ≥98.0% (by GC, titration analysis)
4'-Methylbiphenyl-2-carboxylic acid
Formule: C₁₄H₁₂O₂
MDL Nummer: MFCD00045826
CAS nummer: 7148-03-0
EINECS: 230-462-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fad13865c899faf3677343dc5ec427fd785a11aa&vid=fad13865c899faf3677343dc5ec427fd785a11aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR