5-Chloro-2-nitrotoluene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C2586-5G C2586-25G
TCIAC2586-5GEA 26.4 EUR
TCIAC2586-5G TCIAC2586-25G
5-Chloro-2-nitrotoluene ≥97.0% (by GC)
5-Chloro-2-nitrotoluene
Formule: C₇H₆ClNO₂
Kookpunt: 72°C
Smeltpunt: 25 °C
MDL Nummer: MFCD00024579
CAS nummer: 5367-28-2
EINECS: 226-355-3
UN: 3457
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a52e3ce0ac6d7f1f265a4a85100786ca6fc4d142&vid=e8ef42565d812ceda444a2bed24587423dd9936d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to lump
Purity (GC) min. 97.0 %