D-(-)-Quinic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid
Q0009-25G
Q0009-5G
TCIAQ0009-25GEA
128
EUR
TCIAQ0009-25G
TCIAQ0009-5G
D-(-)-Quinic acid ≥98.0% (by titrimetric analysis)
D-(-)-Quinic acid
Formule:
C₇H₁₂O₆ MW: 192.17 g/mol |
MDL Nummer:
MFCD00003864 CAS nummer: 77-95-2 EINECS: 201-072-8 Merck Index: 14,08059 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -42.0 to -45.0 deg (C=10, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...