(±)-Allyl-2,3-epoxypropyl ether ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Allyl glycidyl ether
A0221-25ML A0221-500ML
TCIAA0221-25MLEA 19.8 EUR
TCIAA0221-25ML TCIAA0221-500ML
(±)-Allyl-2,3-epoxypropyl ether ≥99.0% (by GC)
(±)-Allyl-2,3-epoxypropyl ether
Formule: C₆H₁₀O₂
Kookpunt: 154°C
Vlampunt: 49°C
MDL Nummer: MFCD00005143
CAS nummer: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3df898c00336af12568e8b7c6d08077b578c1f19&vid=3df898c00336af12568e8b7c6d08077b578c1f19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %