4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% (by GC) free radical
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine
A1343-1G A1343-5G
TCIAA1343-1GEA 60.5 EUR
TCIAA1343-1G TCIAA1343-5G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% (by GC) free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Formule: C₉H₁₉N₂O
MW: 171.26 g/mol
Kookpunt: 98°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006479
CAS nummer: 14691-88-4
EINECS: 238-738-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b99ab50e7cd6146706d8e848a4ed4aa34d9fbf&vid=26a8e786d68ae1b52b43c79fbec8d1da7c03fb76&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Orange to Brown powder to lump
Purity (GC) min. 97.0 %
Freezing point 36.0 to 42.0 deg-C