Titanium standard solution, 1000 mg/L Ti in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Leverancier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C061.5N05FP.L1 C061.5N05FPL25 C061.5N05FP.L5
ACSDC061.5N05FP.L1EA 84.9 EUR
ACSDC061.5N05FP.L1 ACSDC061.5N05FPL25 ACSDC061.5N05FP.L5
Titanium standard solution, 1000 mg/L Ti in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Standaarden Kationische standaarden Titanium standaarden