2-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C2386-1G C2386-5G
TCIAC2386-1GEA 26.4 EUR
TCIAC2386-1G TCIAC2386-5G
2-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-methoxyphenylboronic acid
Formule: C₇H₈BClO₃
MDL Nummer: MFCD03411935
CAS nummer: 219735-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4c1308810576c4bfa1abeafd515fd7f4fd5bd50&vid=c4c1308810576c4bfa1abeafd515fd7f4fd5bd50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR