1-Vinyl-2-pyrrolidone 99% stabilised
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone
Stabilisatie: Stabilised with sodium hydroxide
140920050 140925000 140920025
ACRO140920050EA 67.7 EUR
ACRO140920050 ACRO140925000 ACRO140920025
1-Vinyl-2-pyrrolidone 99% stabilised
1-Vinyl-2-pyrrolidone
Formule: C₆H₉NO
Kookpunt: 218 °C (760 mmHg)
Smeltpunt: 13…14 °C
Dichtheid: 1,043 g/cm³
Vlampunt: 95 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003197
CAS nummer: 88-12-0
EINECS: 201-800-4
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5140 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≤0.2 % (Pyrrolidone)
Stabilizer 0.1 % Flaked caustic soda