(-)-Dimethyl-L-malate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (S)-(-)-Dimethyl Malate, (S)-Dimethyl 2-hydroxysuccinate, Dimethyl (S)-(-)-malate

M1343-25G M1343-5G
TCIAM1343-25GEA 60.5 EUR
TCIAM1343-25G TCIAM1343-5G
(-)-Dimethyl-L-malate ≥98.0% (by GC)
(-)-Dimethyl-L-malate
Formule: C₆H₁₀O₅
MDL Nummer: MFCD00066215
CAS nummer: 617-55-0
EINECS: 432-310-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0da672260fc8f3f9661be165babf42a47dac436d&vid=0da672260fc8f3f9661be165babf42a47dac436d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR