Antimony standard solution, 1000 mg/L Sb in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Leverancier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C050.5N1FP.L1 C050.5N1FP.L25 C050.5N1FP.L5
ACSDC050.5N1FP.L1EA 84.9 EUR
ACSDC050.5N1FP.L1 ACSDC050.5N1FP.L25 ACSDC050.5N1FP.L5
Antimony standard solution, 1000 mg/L Sb in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
Standaarden Kationische standaarden Antimonium standaarden