2-Chloro-p-phenylenediamine sulphate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Chlorobenzene-1,4-diamine sulphate, 2-Chloro-1,4-phenylenediammonium sulphate

C0256-25G C0256-500G
TCIAC0256-25GEA 57.2 EUR
TCIAC0256-25G TCIAC0256-500G
2-Chloro-p-phenylenediamine sulphate ≥98.0% (by HPLC, total nitrogen)
2-Chloro-p-phenylenediamine sulphate
Formule: C₆H₉ClN₂O₄S
MDL Nummer: MFCD00013004
CAS nummer: 61702-44-1
EINECS: 262-915-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a027162a78bbb9f5b98fc384dc3c9a6fcb2f9929&vid=a027162a78bbb9f5b98fc384dc3c9a6fcb2f9929&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR