Dicyclohexano-18-crown-6 (mixture of isomers) ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D1668-1G D1668-5G
TCIAD1668-1GEA 31.9 EUR
TCIAD1668-1G TCIAD1668-5G
Dicyclohexano-18-crown-6 (mixture of isomers) ≥98.0% (by GC)
Dicyclohexano-18-crown-6 (mixture of isomers)
Formule: C₂₀H₃₆O₆
Kookpunt: 344°C
MDL Nummer: MFCD00005099
CAS nummer: 16069-36-6
EINECS: 240-216-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 % (total of isomers)
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR