2-(Hydroxymethyl)-18-crown-6 ≥93.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H0990-1G
H0990-200MG
TCIAH0990-1GEA
887
EUR
TCIAH0990-1G
TCIAH0990-200MG
2-(Hydroxymethyl)-18-crown-6 ≥93.0% (by GC)
2-(Hydroxymethyl)-18-crown-6
Formule:
C₁₃H₂₆O₇ Kookpunt: 208°C |
MDL Nummer:
MFCD00188046 CAS nummer: 70069-04-4 EINECS: 274-299-3 |
|
Specification Test Results
Purity (GC) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...