HPTLC plates, nano silica gel 60, modified layers, Nano-SIL CN
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
818184 811115 811116
552-0324EA 228 EUR
552-0324 552-0256 552-0257
HPTLC plates, nano silica gel 60, modified layers, Nano-SIL CN
TLC-platen
Cyano-modified HPTLC silica layers available as glass plates or ALUGRAM® aluminium sheets recommended for steroid hormones, phenols and preservatives.
  • NP or RP separation modes depending on the polarity of the developing solvent
Base material: Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 µm; pH stability 2 to 8.
Modification: Cyanopropyl, carbon content 5,5%
Indicator: Acid-resistant product with a pale blue fluorescence for short-wave UV (254 nm), and UV-absorbing substances appear asMore Product Information
Order Now
Specificaties
Environmentally Preferable