Micro pipettes, disposable, end-to-end
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40551006 40551027
HECH40551006EA 12.8 EUR
HECH40551006 HECH40551027
Micro pipettes, disposable, end-to-end
Pipetten Micro-capillairen
These disposable micro capillaries are calibrated to contain from end to end.
  • With capillary holder (short tube with rubber teat)
  • Filling and discharging can be effected with or without capillary holder
  • Accuracy ≤ ±0,5 %
Verpakking: 250 pieces per pack.