Gel permeation chromatography columns, PLgel Olexis™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1110-1400 PL1110-6400
VARIPL1110-1400EA 629 EUR
VARIPL1110-1400 VARIPL1110-6400
Gel permeation chromatography columns, PLgel Olexis™
Kolommen voor chromatografie
PLgel Olexis is optimized for the analysis of high molecular weight polymers at elevated temperatures, particularly polyolefins and high performance polymers.
  • Optimized design for analysis of polyolefins and high performance polymers
  • Fewer replacements, delays, and failed runs due to column degradation
  • Operates at temperatures up to 220 °C
The column is packed with 13 µm particles designed to strike the best balance between efficiency, lifetime, and resolution without the risk of shear degradation of samples.