Gel permeation chromatography columns, OligoPore™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1113-6520 PL1113-1320 PL1513-1320 PL1213-6520
VARIPL1113-6520EA 2550 EUR
VARIPL1113-6520 VARIPL1113-1320 VARIPL1513-1320 VARIPL1213-6520
Gel permeation chromatography columns, OligoPore™
Kolommen voor chromatografie
OligoPore columns have been developed from an innovative new media that exhibits significantly increased pore volumes compared to conventional low pore size GPC columns. The 300×25 mm preparative column offers high resolution at greatly increased loading for effective isolation of individual components.
  • Near linear calibration curve for best accuracy and precision
  • Very stable media allows for a wide choice of solvents
  • Isolation of individual fractions reveals more information from whole samples
Oligomer fractions collected from the OligoPore preparative column can then be re-injected on analytical columns to check for the purity of the fractions and for comparison with the whole sample. The outcome is higher resolution in the oligomeric region.