High temperature columns, VF-5ht and VF-5ht Ultimetal
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP9047 CP9045 CP9046 CP9048 CP9044 CP9090 CP9091 CP9092 CP9093 CP9094 CP9094I5 CP9095 CP9096 CP9097
VARICP9047EA 573 EUR
VARICP9047 VARICP9045 VARICP9046 VARICP9048 VARICP9044 VARICP9090 VARICP9091 VARICP9092 VARICP9093 VARICP9094 VARICP9094I5 VARICP9095 VARICP9096 VARICP9097
High temperature columns, VF-5ht and VF-5ht Ultimetal
Kolommen voor chromatografie
The VF-5ht improves the analysis of high boiling compounds by exhibiting low bleed, even at high temperatures. Based on ultra-low bleed FactorFour technology, VF-5ht provides unmatched selectivity, sensitivity, and accuracy for the analysis of high-molecular weight compounds.
  • High molecular weight selectivity extends the range of applications
  • Enhanced stability improves column longevity and reduces downtime
  • Superior detector performance gives better detection limits and greater accuracy
VF-5ht is well suited for the separation of non-polar to mid-polar compounds. UltiMetal technology renders the stainless steel inert and enhances the bonding of the stationary phase. The result is long column lifetime with excellent peak shape and low column bleed for the best detection limits at high temperatures, and the lowest cost per analysis.