Capillary columns, CP-Sil 8 CB for pesticides
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7481 CP7504
VARICP7481EA 1440 EUR
VARICP7481 VARICP7504
Capillary columns, CP-Sil 8 CB for pesticides
Kolommen voor chromatografie
CP-Sil 8 CB delivers a linear column response down to femtogram levels.
  • Linear column response down to femtogram levels improves productivity
  • Maximum inertness – tested with DDTs to provide very reliable data
  • Can be used with on-column injection techniques for best detection limits
The column is supplied with a retention gap to avoid problems with solvent condensation, thus allowing repeated splitless injections without phase deterioration. In addition, because of the integrated retention gap, there is no leakage from coupling devices, thereby considerably extending column life.