Capillary columns, CP-Select 624 CB
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7411 CP7414 CP7416 CP7412 CP7415 CP7416I5 CP7417 CP7418 CP7413
VARICP7411EA 792 EUR
VARICP7411 VARICP7414 VARICP7416 VARICP7412 VARICP7415 VARICP7416I5 VARICP7417 VARICP7418 VARICP7413
Capillary columns, CP-Select 624 CB
Kolommen voor chromatografie
The CP-Select 624 CB is a highly reproducible version of the popular 624 phase and has 2-3 times lower bleed than conventional columns.
  • Guaranteed for EPA volatiles with methods 524.2, 624 and 8015 for maximum confidence
  • Excellent peak shape for polar and basic compounds for accurate results
  • Specified by Pharmacopoeia Method V.3.3.9 for residual solvent analysis, providing compliancy
The Select 624 CB, a 6% cyanopropylphenyl, 94% dimethylsiloxane phase is synthesized and fully characterized to ensure maximum column-to-column reproducibility.