TLC plates, silica gel 60, modified layers, RP-2/UV₂₅₄
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
818171 811081 811082
552-0319EA 301 EUR
552-0319 552-0249 552-0250
TLC plates, silica gel 60, modified layers, RP-2/UV₂₅₄
TLC-platen
Dimethyl-modified standard silica layers available as glass plates or ALUGRAM® aluminium sheets. Recommended for active plant constituents and steroids.
  • Silanised silica
  • Normal phase or reversed phase separation modes with purely organic, organic-aqueous or purely aqueous eluents
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm; pH stability 2 to 10.
Modification: Dimethyl, carbon content 4%.
Indicator: Acid-resistant product with a pale blue fluorescence for short-wave UV (254 nm) and UV-More Product Information