Repetitive microdispensing pipette, Acura® 865
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
865.0050 865.0200 865.1000
SOCO865.0050EA 565 EUR
SOCO865.0050 SOCO865.0200 SOCO865.1000
Repetitive microdispensing pipette, Acura® 865
Pipetten Repetitieve pipetten
Handheld microdispenser intended for repeated distribution of microliter volumes. Three models cover the range from 5 to 1000 µl.
  • Excellent ergonomics
  • Ultra soft plunger activation, large volume display
  • Versatile feeding through bottle, tubing or syringe
  • Easy in-lab calibration
  • Autoclavable fully assembled at 121°C
  • Three-way valve system for automatic self-refilling
  • Carefully selected materials ensure durability and optimal resistance to aggressive media
Individually serial numbered and Q.C. certified
Leveringsinformatie: Supplied with 90 cm silicone tubing, stainless steel blunt end cannula, inlet valve cap, Q.C. certificate and operating instructions.