Vials, CrossLab
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8010-0158 8010-0159 8010-0160 8010-0161 8010-0162 8010-0193 8010-0194
HEWL8010-0158EA 0 EUR
HEWL8010-0158 HEWL8010-0159 HEWL8010-0160 HEWL8010-0161 HEWL8010-0162 HEWL8010-0193 HEWL8010-0194
Vials, CrossLab
flesjes
These wide opening translucent vials are made from virgin PP and can be used with crimp or snap caps.
Certificaten: Meets the requirements of 21 CFR 177.1520