Capillary columns, CP-SimDist UltiMetal
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7542 CP7582 CP7592 CP7532 CP7532I5 CP7512 CP7512I5 CP7571 CP7572 CP7570 CP7569I5 CP7562 CP7569 CP7593 CP6540 CP6550
VARICP7542EA 1040 EUR
VARICP7542 VARICP7582 VARICP7592 VARICP7532 VARICP7532I5 VARICP7512 VARICP7512I5 VARICP7571 VARICP7572 VARICP7570 VARICP7569I5 VARICP7562 VARICP7569 VARICP7593 VARICP6540 VARICP6550
Capillary columns, CP-SimDist UltiMetal
Kolommen voor chromatografie
Lower bleed rate than Fused Silica, best column lifetime and accurate results.
  • Conforms to ASTM D 2887 and the extended D 2887 method for compliancy
  • Extended analysis to C120, with maximum temperature of 450 °C
  • UltiMetal tubing for extreme durability