Capillary columns, CP-Sil 5 CB for Formaldehyde
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7475 CP7475I5
VARICP7475EA 1550 EUR
VARICP7475 VARICP7475I5
Capillary columns, CP-Sil 5 CB for Formaldehyde
Kolommen voor chromatografie
CP-Sil 5 CB for Formaldehyde is optimized for the analysis of formaldehyde, water and methanol, and can deliver trace analysis of sulfur compounds and partial permanent gases.
  • Trace analysis of sulfur compounds possible
  • Partial permanent gas analysis possible (especially in switching systems)
  • Non-polar phase provides accurate separations based on volatility
  • High inertness, elutes sulfur components without absorption for high quality data and low detection limits
  • Highest efficiency for this apolar column with the thickest film