PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7574 CP7579 CP7580 CP7581 CP7583 CP7584
VARICP7574EA 852 EUR
VARICP7574 VARICP7579 VARICP7580 VARICP7581 VARICP7583 VARICP7584
PLOT columns, CP-PoraPLOT U and CP-PoraPLOT S
Kolommen voor chromatografie
CP-PoraPLOT U and CP-PoraPLOT S provide symmetrical peaks from polar and non-polar volatiles for ultimate accuracy.
  • Minimal particle loss reduces detector spiking for reliable results
  • Repeatable retention times for better long-term stability
CP-PoraPLOT U provides symmetrical peaks with polar volatiles. Water has no effect on retention times and elutes as a sharp and quantifiable peak. CP-PoraPLOT U is the most polar porous polymer PLOT column and is designed for halogenated compounds, hydrocarbons C1-C6, ketones and solvents.
CP-PoraPLOT S is aMore Product Information