Screw caps for screw neck vials, N 8
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70249 70245 70250 702067 702437 702068 702066 702069
218-1270EA 5.4 EUR
218-1270 MANA70245 MANA70250 MANA702067 MANA702437 MANA702068 MANA702066 MANA702069
Screw caps for screw neck vials, N 8
Sluitingen Sluitingen voor chromatografievials
The suitable screw closures are ready assembled.
  • Septa made from economical Red Rubber/FEP, high quality silicone/PTFE or from virgin PTFE
  • Silicone/PTFE septa are also slitted