Verwarmingsblok, SmartBlock™
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5366000.021 5307000.000 5309000.007 5360000.038 5361000.031 5362000.035 5363000.039 5306000.006 5365000.028 5364000.024 5310000.002 5316000.004 5318000.001
460-0229EA 671 EUR
460-0229 460-0235 460-0221 460-0222 460-0227 460-0228 460-0233 460-0234 460-0230 460-0232 460-0359 460-0358 460-0366
Verwarmingsblok, SmartBlock™
Blokverwarmers Blokken
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
  • Optimised block design supports optimal temperature transfer to sample to achieve excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
  • Automatic block recognition for safe handling
  • Optimised block design ensures maximum temperature transfer to the sample
  • Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.