Specimen containers with lid, SecurTainer™ II
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C576-40MA C576-120MA C576-60MA C576-90MA
SIMPC576-40MAEA 314 EUR
SIMPC576-40MA SIMPC576-120MA SIMPC576-60MA SIMPC576-90MA
Specimen containers with lid, SecurTainer™ II
Monsternamepotjes Containers voor algemeen gebruik
Made of virgin PP and with closures of PE.
  • Uniquely stackable, shatter resistant
  • Straight sided containers are of many sizes
  • Once screwing the cap on the container and the tamper evident sealing ring is automatically locked in place
  • Functional temperature range of –90 to 100 °C
  • Non sterile
Tamper evident containers for collection, transport and storage of specimens.
Order Now
Specificaties
Environmentally Preferable