Gel permeation chromatography columns, EnviroPrep
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1E10-3120EPA PL1E10-6120EPA PL1210-3120EPA PL1210-6120EPA
VARIPL1E10-3120EPAEA 2380 EUR
VARIPL1E10-3120EPA VARIPL1E10-6120EPA VARIPL1210-3120EPA VARIPL1210-6120EPA
Gel permeation chromatography columns, EnviroPrep
Kolommen voor chromatografie
These columns permit a simple, one stage clean-up as part of a methodology to determine pesticides in many organic matrices. The higher molecular weight fractions such as lipids, polymers, natural resins and dispersed high molecular weight components are easily eliminated in the GPC analysis.
  • High sample loading ensures effective trace analysis
  • Simple clean-up procedure saves sample preparation costs
  • Optimised particle size distribution provides high resolution
Preparative GPC for soil extract clean-up is described in EPA Method 3640A using 300×25 mmMore Product Information