Capillary columns, Select Al₂O₃ MAPD
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7431 CP7432 CP7433
VARICP7431EA 1790 EUR
VARICP7431 VARICP7432 VARICP7433
Capillary columns, Select Al₂O₃ MAPD
Kolommen voor chromatografie
The select Al₂O₃ MAPD is an aluminum oxide PLOT column for the analysis of reactive hydrocarbons and is especially targeted towards the response for methyl acetylene and propadiene (MAPD).
  • Two-fold improvement of sensitivity for MAPD decreases detection limits
  • Faster run times improve operating efficiency
  • Improved responses from polar hydrocarbons for better data accuracy
  • Stable up to 200 °C for hydrocarbons
  • Delivers up to a two-fold higher response for MAPD